Would you like to log in again?
SELECT-SAT NT1-77™ Solvent-Ready Non-Woven Cleanroom Wiper, High-Tech Conversions
Supplier: High-Tech Conversions
SS-NT1-77
10141-996CS 264.43 USD InStock
10141-996
SELECT-SAT NT1-77™ Solvent-Ready Non-Woven Cleanroom Wiper, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
SELECT-SAT NT1-77™ dry cleanroom wipers are cleanroom packaged and ready to be saturated with the solvent of your choice.
  • Lint free
  • Solvent ready
  • Cleanroom packaged
  • Non-woven poly-cellulose
  • Convenient clamshell tub
They are comprised of virgin polyester and cellulosic fibers and are virtually lint-free and chemically pure. These wipers are clean-cut and have no nubs or long strands of loose fibers that can be found on perforated roll-type wipes.
Ordering information: Wipers are packaged 100 wipers per unit with 12 units per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty