Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Insulin Receptor beta Mouse Monoclonal Antibody [clone: C18C4]

Supplier: Enzo Life Sciences
905-683-100 905-683-100
101410-452EA 407 USD InStock
CA101410-452 101410-452
Anti-Insulin Receptor beta Mouse Monoclonal Antibody [clone: C18C4]
Antibodies
The Insulin Receptor (InsR) is a heterodimeric receptor tyrosine kinase with an extracellular alpha-chain, a transmembrane domain and an intracellular beta-chain. InsR is activated upon binding of the peptide hormone insulin, leading to autophosphorylation of tyrosine residues 1146, 1150, and 1151 in the activation loop of the beta-chain. Additional autophosphorylation sites such as tyrosine residues 960, 972, 1316, and 1322 regulate the assembly of signal transduction complexes.

Type: Primary
Antigen: Insulin Receptor beta
Clonality: Monoclonal
Clone: C18C4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Sheep, Pig, Bovine, Hamster, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty