Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SMAC/DIABLO Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
905-684-100 905-684-100
101410-454EA 338 USD InStock
CA101410-454 101410-454
Anti-SMAC/DIABLO Mouse Monoclonal Antibody
Antibodies
Smac/DIABLO is one of the pro-apoptotic proteins normally found in the intermembrane space of the mitochondria. In response to apoptotic stimuli, Smac/DIABLO is released into the cytosol resulting in permeabilization of the mitochondrial membrane. Cytosolic Smac/DIABLO activates caspase-9 by binding IAP proteins. Changes in the expression level of Smac/DIABLO have been demonstrated in a number of human cancers including melanoma, ovary, and prostate.

  • For Immunohistochemistry, Western Blot

Recognizes human and monkey Smac/DIABLO.

Type: Primary
Antigen: SMAC/DIABLO
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty