Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AURKB Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-CC203D KAP-CC203-D KAP-CC203F KAP-CC203-F
101410-528EA 309 USD InStock
CA101410-530 101410-528 CA101410-528 101410-530
Anti-AURKB Rabbit Polyclonal Antibody
Antibodies
Aurora A, B, and C have emerged as key mitotic regulators of mammalian centrosome function, chromosome segregation and cytokinesis. Aurora B is one of several chromosomal passenger proteins involved in chromosome alignment at the metaphase plate and cytokinesis. Phosphorylation of a threonine residue in the Aurora activation loop (Thr232 in human Aurora B) is believed to be an essential regulatory mechanism in Aurora kinase activation.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse (weak), rat (weak), bovine (weak) and pig (weak) Aurora B.

Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty