Would you like to log in again?
UltraBed Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14310
101410-644EA 498.57 USD InStock
101410-644
UltraBed Kit, Electron Microscopy Sciences
Stains Microscopy and staining reagents
The kit consists of two components, equal amount, very low viscosity, (~65 cps), in convenient dispenser top bottles. To prepare the embedding resin, just mix equal amounts of the two components. The resin readily infiltrates into specimen and polymerizes to a clear, hard block overnight at 60°C.
  • A Newly Introduced Low Viscosity Epoxy Embedding Kit
  • Mixed resin can be used for both infiltration and embedding
  • All components are shipped together as Non- Hazardous substances
Ordering information: Kit comes with 100 mL each of component A and component B.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty