Would you like to log in again?
Technovit® 4000 Embedding Medium, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Technovit®
14656-02 14656-03
101410-648EA 386.93 USD InStock
101410-648 101410-650
Technovit® 4000 Embedding Medium, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Technovit® 4000 is the low shrinkage embedding medium
It is a fast curing, cold polymerizing, three-component resin, which is based on a modified polyester. It is available in the form of powder, syrup I, and syrup II mixed at a ratio of 2:2:1 (powder:syrup I:syrup II). Syrup I and Syrup II are mixed together first and the powder is mixed in last. The color is white opaque. It shows an excellent margin fit and an optimum grinding and polishing properties.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty