Would you like to log in again?
Histomount, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Histomount
17995-01 17995-05
101410-714EA 91.01 USD InStock
101410-714 101410-716
Histomount, Electron Microscopy Science
Stains Microscopy and staining reagents
A mixture of xylene - mixed isomers and non hazardous ingredients to offer a very good synthetic mountant. Once dried the refractive index is 1.58.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty