Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 1% in aqueous solution

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

21172-1
101410-798EA 43.82 USD InStock
101410-798
Sodium hydroxide 1% in aqueous solution
Sodium hydroxide
  • Sodium hydroxide solution is available in concentrations of 1.0%, 2.5%, 3.1% and 10.0% aqueous


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/00/28984900.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty