Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Jenner's stain solution stain for hematology

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Eosin−Methylene Blue

26033-05
101410-898EA 86.88 USD InStock
101410-898
Jenner's stain solution stain for hematology
Jenner's stain
Jenner Stain solution is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology.

  • High quality staining solution
  • Ready-to-use


Flash Pt: 11 °C (51.8 °F)
Storage Temperature: Ambient
MDL Number: MFCD00081733
CAS Number: 62851-42-7
EINECS: 200-659-6
UN: 1230
ADR: 3,6.1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty