Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Modification of Mayer's Mucihematein, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
26321-01
101411-232EA 92.35 USD InStock
101411-232
Modification of Mayer's Mucihematein, Electron Microscopy Science
Stains Histology Stains
For stain mucins, especially those derived from epithelial cells of glandular tissue.
Lillie (1965); Laskey (1950)
References:
Laskey, Alice: Stain Technology, 25:33 (1959)
Clark G.: Staining Procedures, Williams & Wilkins Co., Baltimore, 3rd Ed., c. 1973, p. 57.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty