Would you like to log in again?
Van Gieson’s Method for Collagen Fibers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26350-01 26350-02 26350-03
101411-250EA 55.6 USD InStock
101411-250 101411-248 101411-246
Van Gieson’s Method for Collagen Fibers, Electron Microscopy Sciences
Stains Histology Stains
Van Gieson’s method for collagen fibers for collagen, muscle and cornified epithelium.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty