Would you like to log in again?
Lillie Modification of Masson’s Trichrome, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
26364-02 26364-03 26364-08 26364-01 26364-04 26364-05 26364-7A 26364-06 26364-07
101411-276EA 35.48 USD InStock
101411-276 101411-266 101411-274 101411-264 101411-272 101411-262 101411-270 101411-260 101411-268
Lillie Modification of Masson’s Trichrome, Electron Microscopy Sciences
Stains Histology Stains
Lillie Modification reagent of Masson’s Trichrome
Lillie (1940)
Stains cells, cytoplasm muscle and collagen of mammalian tissue
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty