Would you like to log in again?
Masson's Trichrome for Connective Tissue, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
26367-02 26367-07 26367-06 26367-05 26367-01 26367-03 26367-04
101411-288EA 108.9 USD InStock
101411-288 101411-286 101411-284 101411-282 101411-290 101411-280 101411-278
Masson's Trichrome for Connective Tissue, Electron Microscopy Sciences
Stains Histology Stains
Masson’s Trichrome for Connective Tissue, Sheehan and Hrapchak (1980), for staining nuclei, agentaffen granules, collagen, cytoplasm, keratin, etc
Stain results - Nuclei: Black, Cytoplasm, Keratin, Muscle Fibers: Red, Collagen, Mucin: Blue.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty