Would you like to log in again?
Weigert's Resorcin Fuchsin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
26370-01 26370-03 26370-04 26370-02 26370-05
101411-298EA 63.75 USD InStock
101411-298 101411-296 101411-294 101411-292 101411-300
Weigert's Resorcin Fuchsin, Electron Microscopy Sciences
Stains Histology Stains
For staining elastic fibers in blood vessel walls, nuclei and collagen.
  • Elastic Tissues: Dark blue
  • Nuclei (red if Orth's Carmine used): Dark blue
  • Collagen: Red-Pink
  • Miscellaneous Tissue Elements: Yellow (with Van Gieson's stain)
Certifications: Certificates of analysis are available.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty