Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Verhoeff’s Van-Gieson’s (VVG Method), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26374-03 26374-06 26374-05 26374-02 26374-SP 26374-01
101411-310EA 40.51 USD InStock
101411-310 101411-308 101411-306 101411-304 101411-302 101411-312
Verhoeff’s Van-Gieson’s (VVG Method), Electron Microscopy Sciences
Stains Histology Stains
Verhoeff’s Van-Gieson’s (VVG Method) is for staining elastic fibers, nuclei, collagen & other tissue elements
Results - Elastic Fibers: Blue-black to Black–Fine elastic fibrils may not be stained with this method, Nuclei: Blue to Black, Collagen: Red
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty