Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Carstairs Method for Fibrin and Platelets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26381-05 26381-02 26381-03 26381-06 26381-04 26381-01
101411-320EA 51.88 USD InStock
101411-320 101411-318 101411-316 101411-324 101411-314 101411-322
Carstairs Method for Fibrin and Platelets, Electron Microscopy Sciences
Stains Hematology Stains
Carstairs Method for Fibrin and Platelets is used for the staining of fibrin and platelets
Stain results - 48 hours or more: Fibrin: Bright Red, Platelets Gray blue to navy, Collagen: Bright Blue, Muscle: Red, Red Blood Cells: Clear Yellow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty