Would you like to log in again?
Jones Method for Kidney, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
26396-03 26396-07 26396-01 26396-11 26396-12 26396-13 26396-14 26396-02 26396-06 26396-09 26396-04 26396-05
101411-364EA 35.5 USD InStock
101411-364 101411-354 101411-362 101411-352 101411-360 101411-350 101411-370 101411-348 101411-368 101411-358 101411-366 101411-356
Jones Method for Kidney, Electron Microscopy Sciences
Stains Histology Stains
Jones Method for Kidney, Silver Method for the staining of basement membranes, reticulum fibers, collagen, and nuclei. Stain results - Basement membranes, reticulum fibers: Black, Nuclei: Blue, Cytoplasm, collagen, connective tissue: Pink-Orange.
Jones, D.B., Amer. J. Path. 27:99 (1951)
AFIP Manual of Histological Staining Methods, 3rd Ed., Ed. L. Luna: NY: McGraw-Hill Publications, c. 1968, p.97
Jones Method for Kidney, Silver Method for the staining of basement membranes, reticulum fibers, collagen, and nuclei
Stain results - Basement membranes, reticulum fibers: Black, Nuclei: Blue, Cytoplasm, collagen, connective tissue: Pink-Orange.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty