Would you like to log in again?
Dane's Method for Prekeratin, Keratin, and Mucin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26401-02 26401-03 26401-01 26401-04
101411-376EA 72.23 USD InStock
101411-376 101411-374 101411-372 101411-378
Dane's Method for Prekeratin, Keratin, and Mucin, Electron Microscopy Sciences
Stains Histology Stains
Dane’s Method for Prekeratin, Keratin, and Mucin. Keratin stain method for the staining of keratin, prekeratin, acid mucopolysaccharides and nuclei. Stain results: Acid Mucopolysaccharides: Blue, Prekeratin and Keranin: Orange to Red, Nuclei: Brown - Orange
Dane, E.T., and Herman, D.L.: Stain Technology, 38:97, 1963.
AFIP Manual of Histological Staining Methods, 3rd Ed., Ed L. Luna: New York: McGraw Hill Publications, c 1968, p.83.
Dane’s Method for Prekeratin, Keratin, and Mucin
Keratin stain method for the staining of keratin, prekeratin, acid mucopolysaccharides and nuclei. Stain results: Acid Mucopolysaccharides: Blue, Prekeratin and Keranin: Orange to Red, Nuclei: Brown - Orange
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty