Would you like to log in again?
Methenamine Silver Method for Argentaffin Cells, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
26481-05 26481-06 26481-04 26481-01 26481-02 26481-07 26481-03
101411-398EA 38.06 USD InStock
101411-398 101411-396 101411-394 101411-392 101411-390 101411-402 101411-400
Methenamine Silver Method for Argentaffin Cells, Electron Microscopy Sciences
Stains Histology Stains
Methenamine Silver Method for Argentaffin Cells for staining of argentaffin granules of enterochromaffin cells, Results - Argentaffin Cells: Black, Coarse connective tissue of submucosa: Variable amount of blackening.
Clark, G.: Staining Procedures, Williams and Wilkins Co., Baltimore, 3rd Ed., c. 1973, p. 161.
Gomori, G., Arch. Path., 45:48, 1948.
Burtner, H.J. and Lillie, R.D., Stain Techn., 24:225-7, 1949.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty