Would you like to log in again?
Taft’s Method for Nucleic Acid, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26777-02 26777-01
101411-556EA 78.63 USD InStock
101411-556 101411-554
Taft’s Method for Nucleic Acid, Electron Microscopy Sciences
Stains Histology Stains
Taft’s Method for Nucleic Acid (1951), Stain DNA & RNA
Stain results - DNA, Deoxryibonucleic Acid: Blue Green, RNA, Ribonucleic Acid: Bright Red.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty