Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lapping and Polishing Fixtures, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50150-50 50150-45
101411-672EA 6304.44 USD InStock
101411-672 101411-658
Lapping and Polishing Fixtures, Electron Microscopy Sciences
Polishers and Tumblers
Lapping and polishing fixtures are designed to accurately produce polished parallel, tapered or critically oriented samples with minimal sub-surface damage.
  • Tungsten carbide base resists wear and maintains sample planarity
  • Wide base ensures stability thereby minimizing sample damage and edge rounding
  • Wear ring is replaceable on some fixtures, which extends the life of the fixture
  • Micrometer depth control allows precise control over sample thickness
  • Precisely crafted stainless steel construction ensures long life and high precision
A series of specialized fixtures has been developed for lapping and polishing samples up to 6" in diameter, TEM samples and oriented crystals. In addition to size and shape, the fixtures also differ in the type of thickness. They are designed to be used either by hand or with a lapping machine for semi-automatic processing (Model 920 above). Sample mounting fixtures are also designed to be used in conjunction with these fixtures to ensure that the sample is mounted parallel to the mounting block and that a thin, uniform layer of wax exists between the mounting block and the sample.
Accessories information: One Vacuum Mounting Block (Series 50153) is required for the Vacuum Mounting Kit and is not included. A custom pattern must be specified by the customer.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty