Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mounting Wax 52, Electron Microscopy Science

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

50390-52
101411-920EA 119.24 USD InStock
101411-920
Mounting Wax 52, Electron Microscopy Science
Stains Microscopy and staining reagents
MWS052 is a tan colored wax, which is translucent on thin films. Softness makes it most useful with abrasive slurry cutting of materials. Best suited to non-porous surfaces such as glass or polished metal with a reasonably large area. Easily soluble in a variety of solvents.

113 Grams/1 Stick, 1" x 1" x 7.5" long (25 x 25 x 190 mm).
Soft Hardness and a Melting Point of 52°C.
Dilutent: Perchloroethylene

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty