Would you like to log in again?
Mounting Wax 70, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
50390-70
101411-922EA 98.33 USD InStock
101411-922
Mounting Wax 70, Electron Microscopy Science
Stains Microscopy and staining reagents
Mounting Wax 70 is a quartz type sticky, orange colored wax (translucent in thin films) used to hold crystals or other materials while they are being sliced, lapped or polished. Moderate hardness and strength make it useful for bonding small samples in diamond wheel sawing and wire sawing operations. Widely used for holding semiconductors, optical materials, ceramics and metals with medium to large contact areas. Chemical resistant. 100 Gram /Pkg of 29 sticks, .25" dia x 4" long (6mm dia x 100mm). Moderate Hardness and a Melting Point of 70°C. Dilutent: Warm soapy water
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty