Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Boron carbide, powder 35 µm

Supplier: Electron Microscopy Sciences
Warning

50510-35
101411-946EA 105.51 USD InStock
101411-946
Boron carbide, powder 35 µm
Boron carbide
Boron Carbide is one of the hardest materials known, ranking third behind diamond and cubic boron nitride.

  • Extreme hardness, (Knoop 100g: 2900-3580 kg/mm2)
  • Good chemical resistance
  • Good nuclear properties
  • Low density (2.52 g/cm2)

Due to the hardness, boron carbide powder is used as an abrasive in polishing and lapping applications.


Formula: CB₄
MDL Number: MFCD00011520
CAS Number: 12069-32-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty