Would you like to log in again?
Serum Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60306-100 60308-20 60308-100 60306-30 60306-05 60306-20 60306-02 60308-05 60306-10 60308-30 60308-10 60306-50 60308-50
101412-406CS 1281.02 USD InStock
101412-406 101412-408 101412-394 101412-396 101412-390 101412-392 101412-402 101412-404 101412-398 101412-388 101412-410 101412-400 101412-412
Serum Bottles, Electron Microscopy Sciences
Bottles Glass Bottles
Clear bottles are manufactured from '400' borosilicate molded glass. Amber bottles are manufactured from '500' borosilicate molded glass.
Certifications: Meets USP Type I standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty