Would you like to log in again?
Lyophilization Seals, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60322-13 60320-07 60330-13N 60321-07 60321-13 60330-20N 60322-07 60320-13
101412-418PK 35.59 USD InStock
101412-418 101412-424 101412-414 101412-416 101412-420 101412-432 101412-422 101412-434
Lyophilization Seals, Electron Microscopy Sciences
Closures Bottle Closures
The Lyophilization seals come in four styles.
The Center Disc Tear Out style exposes the top of the stopper to the syringe. Natural in color. The Flip-Cap, Center Tear style is easy-to-use. Push upward on plastic button to access stopper or septum. No sharp metal edges. Removal of the Flip Cap Button can be accomplished with one hand, freeing the other for the syringe. The Complete Tear Off style seals can be completely removed and contents poured from the bottle. The Lined Aluminum Seals are ideal for filling or retrieving contents with a syringe and as closures for many instrumentation applications. These seals have a center disc tear-out, seal lined with "PTFE" – Faced natural red rubber.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty