Would you like to log in again?
Tough-Tag Labels On a Roll, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Tough-Tags®
62044-O 62045-G 62045-R 62044-R 62044-G 62045-B 62045-Y 62044-B 62044-Y 62045-O
101412-482EA 63.83 USD InStock
101412-482 101412-472 101412-484 101412-474 101412-480 101412-470 101412-468 101412-486 101412-466 101412-478
Tough-Tag Labels On a Roll, Electron Microscopy Sciences
Labels
Tough-Tags are pre-cut, peel-off labels that accept any marking instrument. They are perfectly sized to fit microcentrifuge tubes and other containers in the laboratory.
  • Compatible with any marking instrument
  • Convenient dispensing box assists labeling
  • Use for 0.5 and 1.5 ml tubes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty