Would you like to log in again?
Modular Block for Dri-Bath, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
63124-07 63124-15 63124-17 63124-19 63124-01 63124-34 63124-30 63124-05 63124-09 63124-26
101412-692EA 396.3 USD InStock
101412-692 101412-708 101412-694 101412-714 101412-704 101412-690 101412-710 101412-700 101412-702 101412-686
Modular Block for Dri-Bath, Electron Microscopy Sciences
Block Heaters Blocks
Fit all our Modular Block Dri-Baths.
  • Anodized coating resists the harshest environments
  • Versatile
  • Solid anodized aluminum blocks provide maximum heat retention
  • All blocks include thermometer well for easily measuring block temperature
  • All blocks are the same size: Blocks measure: 3" (7.6cm) W x 2" (5cm) H x 4" (10.1cm) D
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty