Would you like to log in again?
Modular Blocks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
63130-12 63131-02 63131-05 63129-06 63133-02 63129-02 63132-50 63133-01 63129-01 63129-05 63132-10 63129-07 63129-10
101412-736EA 114.09 USD InStock
101412-736 101412-748 101412-738 101412-760 101412-754 101412-766 101412-744 101412-756 101412-746 101412-750 102094-502 101412-762 101412-764
Modular Blocks, Electron Microscopy Sciences
Block Heaters Blocks
Solid black anodized aluminum blocks for better heat retention.
Block includes thermometer well for monitoring block temperatures. Close contact of tubes to block walls, provides excellent heat transfer. Block puller included with all models. For use with all our brand modular dri-baths. Single block size: 3.75W×2.9D×2" H (9.5×7.4×5.1 cm).
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty