Would you like to log in again?
Magic Wands, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63323-02 63323-01 63323-03
101412-836EA 21.05 USD InStock
101412-836 101412-832 101412-834
Magic Wands, Electron Microscopy Sciences
Stirrers Stirring Rods
Magic wands are ideal for use for electronic high-tech areas to laboratories, from preparation of samples to applying adhesives, using it as a stirring rod, or even dosing samples, etc.
  • PEEK polyetheretherketone reinforced with carbon nano
  • Very hard, rigid, high tensile and flexural strength, very high wear resistance
  • High heat capability (260-300°C), good dimension stability, low thermal linear expansion coefficient
  • Excellent resistance to chemicals and aggressive agents, excellent resistance to thermal aging
  • ESD-safe material 106 Ohm
Typical applications include handling of components in cleaning/chemical/assembly processes also at high temperature (soldering).
Probes are 6" (150 mm) long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty