Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

UltraClean™ Glass Slide, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
63472-UC
101412-842PK 400.55 USD InStock
101412-842
UltraClean™ Glass Slide, Electron Microscopy Sciences
Slides Microarray Slides
UltraClean™ substrate provides users with a pristine glass surface which can be used in any coating process compatible with glass such as silanization and polylysine. Manufactured from the highest quality low fluorescence glass.

  • Slides are available with ES (Enhanced Surface Treatment)
  • ES Surface treatments produce a smaller, more uniform spot size without having to change your protocol
  • Bar Coding ability to print all standard bar code formats
  • Soda-lime silicate glass
  • Flatness is 10µ variation over a 76 mm diagonal interval
  • 60/40 scratch-dig surface quality
  • Edges: plain cut
  • Thickness: 1.00 ±0.02 mm
  • Maximum edge chip: 0.05 mm
  • Printable area: 20×70 mm

UltraClean substrates are manufactured in a precise and controlled process incorporating HEPA filtration. This ensures a clean and consistent surface for optimum performance in microarray applications.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty