Would you like to log in again?
Russell's Zenker Fixative, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
64123-05 64123-06
101412-890EA 79.34 USD InStock
101412-890 101412-888
Russell's Zenker Fixative, Electron Microscopy Science
Stains Microscopy and staining reagents
Russell's Zenker Fixative replacement for Zenker's Stock solution.
  • Use in the same manner as Zenker's stock solution
  • Comparable results to Zenker's Stock solution
  • Contains Zinc Chloride
Due to the hazardous nature of mercury, Zenker’s fluid is no longer available. In place we are offering Russell’s Zenker Fixative, which contains Zinc Chloride. The staining results produced by this fixative are comparable to those of Zenker’s Stock solution. Russell’s Modified Zenker’s is used in the same manner as Zenker’s stock solution.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty