Would you like to log in again?
Fluid Dispenser Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64299-02 64299-01
101412-924EA 45.43 USD InStock
101412-924 101412-922
Fluid Dispenser Bottles, Electron Microscopy Sciences
Bottles Dropping and Dispensing Bottles
These ESD safe polyethylene solvent dispensers are available in three standard sizes.
  • Surface resistivity of 10E7 to 10E9
These dispensers have neither a stopper nor a cap to remove. Just touch the dispensing dish with a brush cloth or applicator and solvent is brought to the surface instantly as needed. Stainless steel valves seal contents in bottle until used. The fluid pumps into the dish and it does not drip back into the bottle, keeping stored liquids uncontaminated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty