Would you like to log in again?
Liquid Nitrogen-Cooled Mortar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64470 64472 64471
100496-230EA 258.66 USD InStock
100496-230 100496-228 101413-004
Liquid Nitrogen-Cooled Mortar, Electron Microscopy Sciences
Mortars and Pestles Mortars
The Liquid Nitrogen-Cooled Mortar is for grinding frozen tissue samples without the hazard of liquid nitrogen droplets flying into personnel.
  • Insulated handle 4" (102 mm) is removable for saving space
  • Housing measures 61/2" (165 mm) in diameter, 41/2" high
  • Three suction cup-feet hold the housing
A stainless steel mortar is suspended by three brackets above a stainless steel bowl which holds the liquid nitrogen. The bowl is nested in a well insulated polyethylene housing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty