Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Stirring Rods, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66201-10 66200-40 66201-20 66200-35 66201-15 66200-30 66200-20 66200-10 66200-25 66200-15 66201-25
101413-032EA 22.67 USD InStock
101413-032 102095-948 102095-958 101413-030 101413-036 102095-954 101413-034 102095-956 102095-950 101413-038 102095-952
Stirring Rods, Electron Microscopy Sciences
Stirrers Stirring Rods
Available in two versions: pure "PTFE" solid rod with tapered end and pure "PTFE" with bendable steel core. The version with steel core can be bent into a permanent shape. Rods are inert and will not scratch, use up to 280°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty