Would you like to log in again?
Platinum Crucibles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
66504-4 66505-2 66504-5 66504-6 66505-14 66501-3 66504-8 66504-15 66505-9 66505-8 66504-13 66505-12 66504-12 66505-13 66504-11 66505-3 66504-7 66500-3 66504-3 66504-9 66504-14 66500-1 66501-1 66505-10 66505-6 66505-7 66500-5 66501-5 66505-4 66505-5 66504-2 66504-10 66505-15 66505-11
101413-062EA 1449.98 USD InStock
101413-062 101413-084 101413-082 101413-060 101413-120 101413-088 101413-066 101413-086 101413-064 101413-124 101413-102 101413-100 101413-122 101413-068 101413-106 101413-126 101413-104 101413-108 101413-090 101413-094 101413-072 101413-092 101413-070 101413-076 101413-098 101413-110 101413-096 101413-074 101413-114 101413-078 101413-112 101413-118 101413-116 101413-080
Platinum Crucibles, Electron Microscopy Sciences
Crucibles
Platinum has excellent resistance to chemical attack by acids and fusion mixtures. This is what has made platinum an essential tool in chemical analysis; flux fusions which are widely used on a routine basis for the preparation of samples for x-ray fluorescence etc. All crucibles are made from platinum with an approximate weight in grams and an approximate in millimeter with a tolerance of ±5% . Lids are sold separately and in single units.
Ordering information: EMS strives to maintain accurate pricing. However, due to fluctuations in precious metal prices, pricing on products containing precious metals is not guaranteed. We will contact you if there is a discrepancy.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty