Would you like to log in again?
PTFE Crucibles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
66509-100 66509-75 66509-25 66509-5
101413-132EA 27.02 USD InStock
101413-132 101413-130 101413-134 101413-128
PTFE Crucibles, Electron Microscopy Sciences
Crucibles
Manufactured from pure "PTFE", chemically inert and usable to 280 °C.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty