Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Plastic 150 ml Tube Top Filter 50 mm Membrane, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67043-45 67043-22
101413-142CS 207.26 USD InStock
101413-142 101413-140
Plastic 150 ml Tube Top Filter 50 mm Membrane, Electron Microscopy Sciences
Filters Bottle Top Filters
These tubes minimize unnecessary transfer by filtering directly into the centrifuge tube.
Ordering information: Each case comes with two centrifuge tube stands. The polypropylene tube is supplied with an individually wrapped cap for storage. Individually packaged, sterile, and certified non-pyrogenic.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty