Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70075-O 70075-FP 70075-P 70075-L 70075-Y 70075-T 70075-FG 70075-A 70075-FY 70075-PE 70075-W 70075-GR 70075-B 70075-G
101413-220PK 441.21 USD InStock
101413-220 101413-212 101413-224 101413-202 101413-210 101413-222 101413-216 101413-228 101413-206 101413-214 101413-226 101413-204 101413-218 101413-208
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Embedding Cassettes Biopsy Processing Cassettes
Biopsy processing cassettes with four square compartments each measuring 13 mm.
  • Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty