Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Disposable Pipettes – Sterile, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70954-5 70953-10 70954-2 70954-1 70954-10 70953-1 70954-5S 70953-5 70953-25 70953-50
101413-242PK 80.25 USD InStock
101413-242 101413-250 101413-256 101413-246 101413-254 101413-244 101413-238 101413-236 101413-258 101413-248
Disposable Pipettes – Sterile, Electron Microscopy Sciences
Pipettes Pasteur/Transfer Pipettes
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to ±1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty