Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Narrow Stem Pipettes - Sedi-Pet, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70966-12 70966-12S 70967-14S 70967-13S 70967-13 70967-14
101413-328PK 129.07 USD InStock
101413-328 101413-330 101413-322 101413-332 101413-326 101413-324
Narrow Stem Pipettes - Sedi-Pet, Electron Microscopy Sciences
Pipettes Pasteur/Transfer Pipettes
These polyethylene disposable have a low-affinity surface, attached bulb avoids cross-contamination, and are one step biohazard disposable. These Micro-tip pipettes are ideal for loading gels, DNA sample removal, protein assays, lipids extractions, drop-wise additions to culture plates, using 2 ml auto sampler vials, remove or disperse small volumes, etc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty