Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Double Edge Razor Blade Platinum Coated, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

72003-01 72003-10
101413-476PK 30.05 USD InStock
101413-476 101413-478
Double Edge Razor Blade Platinum Coated, Electron Microscopy Sciences
Razors and Blades
Made from stainless steel, coated with Platinum.

Super-sharp-edges. Measures: 62.96 x 21.95 x 0.10 mm (1.69 x 0.865 x 0.004").

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty