Would you like to log in again?
Microsurgical Knife, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72047-15 72047-30 72047-45
101413-490PK 91.74 USD InStock
101413-490 101413-488 101413-486
Microsurgical Knife, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpels
Made from tool steel wire. Sharpened to microsurgical quality.
The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
Packaging: 6 blister packs to a box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty