Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dumont Tweezers Style SS, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
72812-DZ 72812-D
101413-564PK 510.14 USD InStock
101413-564 101413-562
Dumont Tweezers Style SS, Electron Microscopy Sciences
Forceps
Dumoxel is an alloy composed of C, Cr, Ni, Mo and Cu. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium.

  • Dumoxel is 95% antimagnetic
  • Resistant against temperatures of around 400 °C (DIN 50 914)
  • Suitable for autoclave sterilization at 270 °C

Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids.

Length: 135 mm.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty