Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Graphite, foil boat, pyrolytic B1

Supplier: Electron Microscopy Sciences

73808-B1
101413-606EA 226.49 USD InStock
101413-606
Graphite, foil boat, pyrolytic B1
Graphite
Boat dimensions: Thickness of pyrolytic graphite foil is 0.51 mm (0.020”). Overall length is 32 mm (1.25”) and width is 9.6 mm (0.375”).
Trough Dimension L x W x D: 12.7 x 9.0 x 3.2 mm (0.5 x 0.355 x 0.03”).


Formula: C
MW: 12.01 g/mol
MDL Number: MFCD00144065
CAS Number: 7782-42-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty