Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Cryogenic Labels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77025-10 77026-11 77027-16 77028-18 77027-01 77026-15 77026-10 77027-17 77025-2A 77027-12 77025-01 77026-17 77027-10 77027-15 77027-11 77026-01 77026-16 77026-12 77025-11
101413-740EA 101.91 USD InStock
101413-740 101413-730 101413-722 101413-744 101413-734 101413-742 101413-720 101413-732 101413-726 101413-748 101413-716 101413-738 101413-746 102098-468 101413-724 101413-736 102098-466 101413-728 101413-718
Cryogenic Labels, Electron Microscopy Sciences
Labels
Cryogenic labels are made from top quality materials with wide range of sizes, designed specially for cryogenic use.
DuraStick Labels are suitable to be used under a temperature of −40 °C. These labels are features with smudge resistant surface and are available in rolls for thermal transfer printing or in 8½×11" sheets for laser printing. All accept hand written notations with cryo markers.
CryoStick Labels are suitable to be used for temperatures of −80 °C. Ideal for use with cryotubes, these labels are best used with thermal transfer printers or cryo marker pens. Available in a wide range of sizes on sheets or rolls, they are not recommended for laser printing.
LiquiStick Labels are suitable to be used for temperatures of −195 °C. Labels feature with the synthetic surface that accepts crisp bar codes from thermal transfer printers or handwritten notations cryo markers. Supplied in sheets or rolls, these labels are not recommended for use with laser printers.
Satin Cloth Labels are suitable to be used for temperatures of −60 °C. These textured labels withstand dry ice, deep freezers and liquid gases. Supplied in sheets or rolls. They are easily marked with cryo markers, not for use with automated printers.
Wrap-Around Labels are suitable for freezers and cryogenic use. These unique labels are for sub-zero use, measures ¹⁵/₁₆×2³/₈". They feature a ¹⁵/₁₆×¹⁵/₁₆" area, frosted for thermal printing or marking with cryo markers. The remainder of the label is clear. It wraps around the cryo-tube to provide a protective cover for printed information while leaving a clear space for viewing the tube contents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty