Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

EMS Fiber Tip Tweezers, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
78016-RF 78094-CFZ 78080-CF 78084-CF 78090-CFZ 78090-CF 78095-CF 78010-RF 78010-CP 78084-CP 78012-RFZ 78092-CP 78095-CP 78085-CF 78010-RFZ 78092-CF 78080-CP 78094-CF 78014-CP 78014-RFZ 78016-CP 78085-CP 78094-CP 78014-RF 78090-CP 78080-CFZ 78012-CP 78012-RF 78092-CFZ
100502-282EA 45.54 USD InStock
100502-282 100502-280 101413-986 100499-836 100499-838 101413-988 100494-598 100499-834 100494-596 100494-594 101414-000 101414-002 101414-004 101414-006 101413-990 100494-592 101413-994 100502-874 101413-992 101413-998 101413-996 100499-842 100494-600 100499-844 102098-814 100502-876 100499-840 100502-878 100494-602
EMS Fiber Tip Tweezers, Electron Microscopy Sciences
Forceps
Tweezers with anti-magnetic steel handles and synthetic fiber tips.

Synthetic Fiber has high-performance Plastic type CP - PEEK/CF30 (Polyetheretherketone reinforced with 30 wt% carbon fiber). Very hard, high tensile and flexural strength. High heat- 260 – 300°C – resistance to thermal aging. Excellent chemical resistance. ESD safe – low surface resistivity (102 Ohm).
Engineering Plastic type CF - PA66/CF30 (Polyamide 66 reinforced with 30 wt% carbon fiber) is heat stable(-40°C to 190°C). Rigidity, excellent tensile strength and flexural strength. Good chemical resistance (oils, greases, fuels, non-polar solvents); not resistant to strong acids, alkalis and hot water or steam. ESD safe, (avoids dust attraction, sparks, ignition sources). Good conductivity.
Non Conductive Plastic RF – PET/GF30 (Polyethylene terephtalate reinforced with 30 wt% glass fiber) has high tensile and flexural strength. Good chemical resistance and low moisture absorption. Electrically insulation. High temperature capability. White in color.

The handles are made from antimagnetic steel and tips from synthetic fiber. Temperature Resistance: °F (190°C).

Style 00: A = 130 mm (5”); B = 1.0 mm (0.04”); C = 2.0 mm (0.08”)
Style 2A: A = 130 mm (5”); B = 1.8 mm (0.07”); C = 1.0 mm (0.04”)
Style 2AB: A = 130 mm (5”); B = 1.8 mm (0.07”; C =1.0 mm (0.04”)
Style 3: A = 130 mm (5”); B = 0.8 mm (0.03”); C = 1.3 mm (0.05”)
Style 5: A = 130 mm (5”); B = 0.5 mm (0.02”); C = 0.6 mm (0.025")
Style 5X: A = 130 mm (5”); B = 0.5 mm (0.02”); C = 0.6 mm (0.025”)
Style 7: A = 130 mm (5”); B = 0.6 mm (0.025"); C = 0.6 mm (0.025”)
Style 259: A = 130 mm (5”); B = 0.6 mm (0.025"); C = 1.0 mm (0.04”)
Style 269: A = 130 mm (5”); B = 3.0 mm (0.12”); C = 1.8 mm (0.07”)
Model 248: A = 130 mm (5”); B = 10 mm (0.4”); C = 1.8 mm (0.07”)
Style 250: A = 130 mm (5”); B = 10 mm (0.4”); C = 1.8 mm (0.07”)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty