Would you like to log in again?
DNA Gel Loading Buffer, Omega Bio-Tek
Supplier: Omega Bio-Tek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AC10098 AC10097 AC10098 AC10088 AC10088 AC10097
101414-298EA 73.92 USD InStock
CA101414-298 101414-298 CA101414-300 101414-300 CA101414-302 101414-302
DNA Gel Loading Buffer, Omega Bio-Tek
Buffers Electrophoresis Buffers Loading Buffers
The 50X TAE buffer is composed of 2M tris-acetate and 0.05M EDTA at pH 8.3
Loading buffers are intended to be mixed with samples containing DNA in order to facilitate loading of the samples into the wells of agarose and acrylamide gels.
It is ideal for preparation of agarose gels for electrophoresis.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty