Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
dam-/dcm- Competent E.coli, New England Biolabs
Supplier: New England Biolabs
C2925I C2925I C2925H C2925H
101417-894EA 214 USD InStock
101417-894 200064-394 CA101417-894 CA200064-394
dam-/dcm- Competent E.coli, New England Biolabs
Competent Cells
Methyltransferase deficient chemically competent E.coli cells suitable for growth of plasmids free of Dam and Dcm methylation.
Note that dam- strains are not recommended as a host for primary cloning/ligation. The dam mutation can result in an increased mutation rate in the cell and a reduction in the transformation efficiency. DNA should be maintained in a dam+ strain unless there is a specific need for DNA free of Dam or Dcm methylation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty