Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NEB®5-alpha Competent E.coli (High Efficiency), New England Biolabs
Supplier: New England Biolabs
NEB®
C2987I C2987H C2987R C2987H C2987U C2987P C2987I C2987R C2987P C2987U
101417-898EA 207 USD InStock
CA102855-190 101417-898 CA101417-900 CA101417-898 103218-958 103218-956 102855-190 CA103218-958 101417-900 CA103218-956
NEB®5-alpha Competent E.coli (High Efficiency), New England Biolabs
Competent Cells
NEB 5-alpha Competent E. coli (High Efficiency) is a derivative of the popular DH5. It is T1 phage resistant and endA deficient for high-quality plasmid preparations.
  • High efficiency strain ideal for a wide variety of applications
  • In a wide variety of sizes, including single-use vials, 200 µl vials, 96- and 384-well plates
  • No dry ice surcharge on competent cell shipments
  • Outgrowth medium included
  • Free of animal products
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty